Please login to your online account to display your discounted pricing

anti-PGP9.5 / UCHL-1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-100 of human UchL1 was used as the immunogen for this antibody.
Antigen PGP9.5 / UCHL-1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224781

 
novus biologicals canada
NBP224781
Unlabeled 0.1mg Each for $255.80

NBP224781SS

 
novus biologicals canada
NBP224781SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-100 of human UchL1 was used as the immunogen for this antibody.
Antigen PGP9.5 / UCHL-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P09936
Gene ID (Entrez) 7345
Formulation 100μg antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL