Please login to your online account to display your discounted pricing

anti-PGP9.5 / UCHL-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11058874SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11058874T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7345
Gene Accession No. P09936
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Immunogen Recombinant full length human UCHL1 purified from E. coli. [UniProt No. P09936]
Format Antiserum
Antigen PGP9.5 / UCHL-1
Host Species Rabbit