missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-PGD2 Synthase/PTGDS, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to region within residue 120-190 of human Prostaglandin D Synthase/Lipocalin. [Swiss-Prot No. P41222]
Antigen PGD2 Synthase/PTGDS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11059911 Novus Biologicals Canada
NB11059911
0.1mL Each for $329.00
NB11059911T Novus Biologicals Canada
NB11059911SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
PGD2 Synthase/PTGDS
Affinity Purified
RUO
Human, feline, bovine, rabbit, canine, rat, mouse and primate. Immunogen has 89% homology to equine, 85% homology to sheep and 82% homology to porcine.
5730
1mg/mL
The epitope recognized by this antibody maps to region within residue 120-190 of human Prostaglandin D Synthase/Lipocalin. [Swiss-Prot No. P41222]
Polyclonal
Primary
Western Blot
B2R727
PBS, 30% glycerol