Please login to your online account to display your discounted pricing

anti-PGD2 Synthase/PTGDS, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to region within residue 25-100 of human Prostaglandin D Synthase/Lipocalin. [Swiss-Prot No. P41222]
Antigen PGD2 Synthase/PTGDS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11059909

 
novus biologicals canada
NB11059909
Unlabeled 0.1mL Each for $329.00

NB11059909T

 
novus biologicals canada
NB11059909SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to region within residue 25-100 of human Prostaglandin D Synthase/Lipocalin. [Swiss-Prot No. P41222]
Antigen PGD2 Synthase/PTGDS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity Human, primate, mouse, rat, bovine, porcine, feline, equine and canine. Immunogen sequence has 87% homology to sheep and 82% homology to rabbit.
Gene Accession No. B2R727
Gene ID (Entrez) 5730
Formulation PBS, 30% glycerol
Concentration 1mg/mL