Please login to your online account to display your discounted pricing

anti-PGC1 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP104676SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP104676SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1.0mg/mL
Test Specificity Human and Mouse. Squirrel reactivity reported in the scientific literature. Expected reactivity based on sequence identity: monkey (98%), equine (94%), canine (93%) and rat (88%). Rat reactivity reported in scientific literature (PMID: 22208735).
Gene ID (Entrez) 10891
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9UBK2
Formulation PBS
Quantity 0.025mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Squirrel
Conjugate Unlabeled
Primary or Secondary Primary
Antigen PGC1 alpha
Regulatory Status RUO
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of the human PGC-1 alpha protein (within residues 400-550). [Swiss-Prot No. Q9UBK2].
Host Species Rabbit