Please login to your online account to display your discounted pricing

anti-PGAM1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PGAM1 protein (between residues 200-254) [UniProt P18669]
Antigen PGAM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149532

 
novus biologicals canada
NBP149532
Unlabeled 0.1mL Each for $329.00

NBP149532SS

 
novus biologicals canada
NBP149532SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PGAM1 protein (between residues 200-254) [UniProt P18669]
Antigen PGAM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Gene ID (Entrez) 5223
Formulation PBS, 30% glycerol
Concentration 1mg/mL