Please login to your online account to display your discounted pricing

anti-PGAM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149532

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149532

  • / Each
 

Description & Specifications

Specifications

Antigen PGAM1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PGAM1 protein (between residues 200-254) [UniProt P18669]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5223
Target Species Human
Target Species Murine