Please login to your online account to display your discounted pricing

anti-Peroxiredoxin 6 Polyclonal, Thermo Scientific™

Peroxiredoxin 6 Polyclonal Ab PA1-18281 from Thermo Scientific™ specifically detects Peroxiredoxin 6 in Hu samples, and it is validated for ELISA, IHC (Paraffin, Frozen) and WB

$506.05 - $506.05

Specifications

Antigen Peroxiredoxin 6
Cross Reactivity Human
Format Serum
Host Species Sheep
Immunogen Recombinant rat Peroxiredoxin-6.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-534

 
pierce biotechnology inc
PA118281
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen Peroxiredoxin 6
Cross Reactivity Human
Format Serum
Host Species Sheep
Immunogen Recombinant rat Peroxiredoxin-6.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Peroxiredoxin 6 Polyclonal Ab PA1-18281 from Thermo Scientific™ specifically detects Peroxiredoxin 6 in Hu samples, and it is validated for ELISA, IHC (Paraffin, Frozen) and WB

Peroxiredoxin-6 is involved in redox regulation of the cell. Can reduce H(2)O(2) and short chain organic, fatty acid, and phospholipid hydroperoxides. Peroxiredoxin-6 may also play a role in the regulation of phospholipid turnover as well as in protection against oxidative injury.
Host Species: Sheep
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant rat Peroxiredoxin-6.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Prx6, NSGPx
Uniprot Accession: P30041
Gene Symbol: PRDX6
Entrez GeneID: 9588
PA1-18281 detects Peroxiredoxin 6 in Human samples. PA1-18281 has been successfully used in ELISA, Immunohistochemistry (Paraffin, Frozen) and Western Blot procedures. PA1-18281 The immunogen is Recombinant rat Peroxiredoxin-6..