Please login to your online account to display your discounted pricing

anti-Periostin/OSF-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130042SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130042SS

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Regulatory Status RUO
Format Affinity Purified
Antigen Periostin/OSF-2
Conjugate Unlabeled
Formulation PBS
Test Specificity Specific for the ∽93 kDa periostin doublet in mouse lung extract. Consistent with the fact that this antibody is made against an alternatively spliced region of periostin, this antibody recognizes only the largest two of the three forms of periostin recognized on Western blots by the Pan periostin antibody and also shows a distinctive staining pattern by immunohistochemistry. The antibody works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90°C in 10mM sodium citrate (pH 6.0)/ 0.1% Tween-20). Expected reactivity to all mammalian and avian species. Based on customer review rabbit reacts.
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Gene ID (Entrez) 10631
Primary or Secondary Primary
Host Species Rabbit
Quantity 0.02mL
Immunogen Bacterial fusion protein equivalent to a 188-amino acid polypeptide from the C-terminal region of mouse periostin which is comprised of six small alternatively-spliced exons.