Please login to your online account to display your discounted pricing

anti-Periostin/OSF-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130042

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130042

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Immunogen Bacterial fusion protein equivalent to a 188-amino acid polypeptide from the C-terminal region of mouse periostin which is comprised of six small alternatively-spliced exons.
Formulation PBS
Gene ID (Entrez) 10631
Concentration 0.5mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Specific for the ∽93 kDa periostin doublet in mouse lung extract. Consistent with the fact that this antibody is made against an alternatively spliced region of periostin, this antibody recognizes only the largest two of the three forms of periostin recognized on Western blots by the Pan periostin antibody and also shows a distinctive staining pattern by immunohistochemistry. The antibody works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90°C in 10mM sodium citrate (pH 6.0)/ 0.1% Tween-20). Expected reactivity to all mammalian and avian species. Based on customer review rabbit reacts.
Conjugate Unlabeled
Quantity 0.1mL
Antigen Periostin/OSF-2
Format Affinity Purified
Regulatory Status RUO