missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Periostin/OSF-2, Polyclonal, Novus Biologicals™

$110.17 - $362.00

Specifications

Host Species Rabbit
Immunogen Bacterial fusion protein equivalent to a 188-amino acid polypeptide from the C-terminal region of mouse periostin which is comprised of six small alternatively-spliced exons.
Antigen Periostin/OSF-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP130042SS Novus Biologicals Canada
NBP130042SS
0.02mL Each for $110.17
NBP130042 Novus Biologicals Canada
NBP130042
0.1mL Each for $362.00
Specifications Description & Specifications

Specifications

Rabbit
Periostin/OSF-2
Affinity Purified
RUO
Specific for the ∽93 kDa periostin doublet in mouse lung extract. Consistent with the fact that this antibody is made against an alternatively spliced region of periostin, this antibody recognizes only the largest two of the three forms of periostin recognized on Western blots by the Pan periostin antibody and also shows a distinctive staining pattern by immunohistochemistry. The antibody works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90°C in 10mM sodium citrate (pH 6.0)/ 0.1% Tween-20). Expected reactivity to all mammalian and avian species. Based on customer review rabbit reacts.
PBS
Bacterial fusion protein equivalent to a 188-amino acid polypeptide from the C-terminal region of mouse periostin which is comprised of six small alternatively-spliced exons.
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
10631
0.5mg/mL