Please login to your online account to display your discounted pricing

anti-Perilipin-3/TIP47, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region within the N-terminus (within residues 1-100) of the mouse TIP47 protein. [Swiss-Prot No. Q9DBG5]
Antigen Perilipin-3/TIP47
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11040765

 
novus biologicals canada
NB11040765
Unlabeled 0.1mL Each for $309.05

NB11040765T

 
novus biologicals canada
NB11040765SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region within the N-terminus (within residues 1-100) of the mouse TIP47 protein. [Swiss-Prot No. Q9DBG5]
Antigen Perilipin-3/TIP47
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Gene ID (Entrez) 10226
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL