Please login to your online account to display your discounted pricing

anti-Perilipin-3/TIP47, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11040765SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11040765T

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Immunogen A synthetic peptide made to a region within the N-terminus (within residues 1-100) of the mouse TIP47 protein. [Swiss-Prot No. Q9DBG5]
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10226
Quantity 0.025mL
Host Species Rabbit
Regulatory Status RUO
Antigen Perilipin-3/TIP47