Please login to your online account to display your discounted pricing

anti-Perilipin-3/TIP47, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11040765

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11040765

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 10226
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Format Affinity Purified
Immunogen A synthetic peptide made to a region within the N-terminus (within residues 1-100) of the mouse TIP47 protein. [Swiss-Prot No. Q9DBG5]
Concentration 1mg/mL
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Antigen Perilipin-3/TIP47
Regulatory Status RUO