Please login to your online account to display your discounted pricing

anti-PER2, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 750-800 of human PER2 was used as the immunogen.
Antigen PER2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224616SS

 
novus biologicals canada
NBP224616SS
Unlabeled 0.025mg Each for $109.00

NBP224616

 
novus biologicals canada
NBP224616
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 750-800 of human PER2 was used as the immunogen.
Antigen PER2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse, 94% homologous in human, monkey and horse, and 77% homologous in dog.
Gene Accession No. O15055
Gene ID (Entrez) 8864
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL