Please login to your online account to display your discounted pricing

anti-PER2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224616SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224616SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse, 94% homologous in human, monkey and horse, and 77% homologous in dog.
Quantity 0.025mg
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen PER2
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O15055
Gene ID (Entrez) 8864
Concentration 0.5mg/mL
Host Species Rabbit
Immunogen A portion of amino acids 750-800 of human PER2 was used as the immunogen.
Regulatory Status RUO