Please login to your online account to display your discounted pricing

anti-PER2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224616

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224616

  • / Each
 

Description & Specifications

Specifications

Antigen PER2
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O15055
Host Species Rabbit
Immunogen A portion of amino acids 750-800 of human PER2 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8864
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse, 94% homologous in human, monkey and horse, and 77% homologous in dog.
Target Species Murine
Target Species Rat