Please login to your online account to display your discounted pricing

anti-PER1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 30-80 of mouse PER1 was used as the immunogen.
Antigen PER1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224692SS

 
novus biologicals canada
NBP224692SS
Unlabeled 0.025mg Each for $109.00

NBP224692

 
novus biologicals canada
NBP224692
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 30-80 of mouse PER1 was used as the immunogen.
Antigen PER1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in strains H1N1, H3N2, H5N1, H6N1, H6N2 and H9N2, and 93% homologous in H7N7.
Gene ID (Entrez) 5187
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL