Please login to your online account to display your discounted pricing

anti-PER1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224692SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224692SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in strains H1N1, H3N2, H5N1, H6N1, H6N2 and H9N2, and 93% homologous in H7N7.
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5187
Concentration 0.5mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.025mg
Immunogen A portion of amino acids 30-80 of mouse PER1 was used as the immunogen.
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Formulation 100μg in 200μL PBS containing 0.05% BSA
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Mouse
Cross Reactivity Porcine
Format Purified
Regulatory Status RUO
Antigen PER1