Please login to your online account to display your discounted pricing

anti-PER1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224692

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224692

  • / Each
 

Description & Specifications

Specifications

Antigen PER1
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Mouse
Cross Reactivity Porcine
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 30-80 of mouse PER1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5187
Test Specificity The amino acid sequence used as immunogen is 100% homologous in strains H1N1, H3N2, H5N1, H6N1, H6N2 and H9N2, and 93% homologous in H7N7.
Target Species Bovine
Target Species Canine
Target Species Equine
Target Species Murine
Target Species Porcine