Please login to your online account to display your discounted pricing

anti-Pellino 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056672

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056672

  • / Each
 

Description & Specifications

Specifications

Antigen Pellino 1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Antisera
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Immunogen Genbank No. NP_065702.2.
Quantity 0.2mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 57162
Target Species Human
Target Species Murine