Please login to your online account to display your discounted pricing

anti-PEDF R/PNPLA2/ATGL, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region (residues 400-500) of human ATGL. [Accession No. NP_005171]
Antigen PEDF R/PNPLA2/ATGL
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11041536T

 
novus biologicals canada
NB11041536SS
Unlabeled 0.025mL Each for $109.00

NB11041536

 
novus biologicals canada
NB11041536
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region (residues 400-500) of human ATGL. [Accession No. NP_005171]
Antigen PEDF R/PNPLA2/ATGL
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. Q96AD5
Gene ID (Entrez) 57104
Formulation Tris-glycine, 150mM NaCl
Concentration 0.76mg/mL