Please login to your online account to display your discounted pricing

anti-PEDF R/PNPLA2/ATGL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11041536SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11041536T

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Primary or Secondary Primary
Antigen PEDF R/PNPLA2/ATGL
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q96AD5
Regulatory Status RUO
Concentration 0.76mg/mL
Conjugate Unlabeled
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 57104
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Immunogen A synthetic peptide within the C-terminal region (residues 400-500) of human ATGL. [Accession No. NP_005171]
Quantity 0.025mL