Please login to your online account to display your discounted pricing

anti-PEDF R/PNPLA2/ATGL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11041536

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11041536

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled
Concentration 0.76mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 57104
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q96AD5
Regulatory Status RUO
Host Species Rabbit
Immunogen A synthetic peptide within the C-terminal region (residues 400-500) of human ATGL. [Accession No. NP_005171]
Format Affinity Purified
Quantity 0.1mL
Antigen PEDF R/PNPLA2/ATGL