Please login to your online account to display your discounted pricing

anti-Pea3, Polyclonal, Novus Biologicals™

$418.00 - $418.00

Specifications

Host Species Rabbit
Immunogen This antibody was made against a protein fragment from the N Terminus Region (NM_001986).
Antigen Pea3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

20580002

 
novus biologicals canada
20580002
Unlabeled 0.05mg Each for $418.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was made against a protein fragment from the N Terminus Region (NM_001986).
Antigen Pea3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This product is specific for Human ETV4.
Gene Accession No. P43268
Gene ID (Entrez) 2118
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0