Please login to your online account to display your discounted pricing

anti-PDX-1/IPF1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222150

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222150

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Test Specificity Human and mouse. Predicted to react with rat based on 100% sequence homology.
Gene Accession No. P52945
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Immunogen A synthetic peptide made to a C-terminal portion of the human PDX1 protein (between residues 100-200) [UniProt P52945]
Formulation PBS
Gene ID (Entrez) 3651
Concentration 1.0mg/mL
Host Species Rabbit
Regulatory Status RUO
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Antigen PDX-1/IPF1