Please login to your online account to display your discounted pricing

anti-PDK4, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot No. Q16654]
Antigen PDK4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP107047

 
novus biologicals canada
NBP107047
Unlabeled 0.1mL Each for $362.00

NBP107047SS

 
novus biologicals canada
NBP107047SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot No. Q16654]
Antigen PDK4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human and Mouse. Predicted to react with bovine based on 100% sequence homology. Immunogen sequence has 94% homology to rat.
Gene Accession No. Q16654
Gene ID (Entrez) 5166
Formulation PBS, 30% glycerol
Concentration 1.27mg/mL