Please login to your online account to display your discounted pricing

anti-PDK4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107047SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107047SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5166
Conjugate Unlabeled
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. Q16654
Applications Western Blotting
Test Specificity Human and Mouse. Predicted to react with bovine based on 100% sequence homology. Immunogen sequence has 94% homology to rat.
Concentration 1.27mg/mL
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot No. Q16654]
Antigen PDK4
Host Species Rabbit