Please login to your online account to display your discounted pricing

anti-PDK4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP107047

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP107047

  • / Each
 

Description & Specifications

Specifications

Antigen PDK4
Applications Western Blot
Concentration 1.27mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q16654
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot No. Q16654]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5166
Test Specificity Human and Mouse. Predicted to react with bovine based on 100% sequence homology. Immunogen sequence has 94% homology to rat.
Target Species Human
Target Species Murine