Please login to your online account to display your discounted pricing

anti-PDK-1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PDK1 protein sequence (between residues 350-436). [Swiss-Prot Q15118]
Antigen PDK-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002383

 
novus biologicals canada
NB1002383
Unlabeled 0.1mL Each for $309.05

NB1002383SS

 
novus biologicals canada
NB1002383SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PDK1 protein sequence (between residues 350-436). [Swiss-Prot Q15118]
Antigen PDK-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human. Predicted to react with mouse and rat based on 100% sequence homology. Immunogen sequence has 93% homology to chicken.
Gene Accession No. Q15118
Gene ID (Entrez) 5163
Formulation Tris-glycine, 150mM NaCl
Concentration 0.4mg/mL