Please login to your online account to display your discounted pricing

anti-PDK-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002384

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002384

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Test Specificity Human. Predicted to react with rat, mouse, primate and bovine based on 100% sequence homology. Immunogen sequence has 92% homology to chicken, 91% homology to Zebrafish and porcine.
Gene ID (Entrez) 5163
Cross Reactivity Human
Gene Accession No. Q15118
Format Affinity Purified
Regulatory Status RUO
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Host Species Rabbit
Primary or Secondary Primary
Antigen PDK-1
Applications Western Blotting
Quantity 0.1mL
Immunogen A synthetic peptide made to an internal portion of the human PDK1 protein sequence (between residues 300-400). [Swiss-Prot Q15118]