Please login to your online account to display your discounted pricing

anti-PDE9A, Polyclonal, Novus Biologicals™

$182.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Sequence from the C-terminal region of PDE9A
Antigen PDE9A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP100641

 
novus biologicals canada
NBP100641
Unlabeled 0.05mL Each for $329.00

NBP100641SS

 
novus biologicals canada
NBP100641SS
Unlabeled 0.025mL Each for $182.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Sequence from the C-terminal region of PDE9A
Antigen PDE9A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene ID (Entrez) 5152
Formulation PBS, 30% glycerol
Concentration 1mg/mL