missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
pre-B-cell leukemia homeobox 1, Mouse, Clone: 4A2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PBX1.
Supplier: Abnova Corporation H00005087M01
Description
Sequence: MQLKQSTCEAVMILRSRFLDARRKRRNFNKQATEILNEYFYSHLSNPYPSEEAKEELAKKCGITVSQVSNWFGNKRIRYKKNIGKFQEEANIYAAKTAVTATNVSAHGSSpecifications
pre-B-cell leukemia homeobox 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002585 | |
PBX1 | |
PBX1 (NP_002576, 213 a.a. ∼ 321 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
4A2 | |
Mouse monoclonal antibody raised against a partial recombinant PBX1. | |
PBX1 | |
DKFZp686B09108/MGC126627 | |
Mouse | |
Affinity Purified | |
RUO | |
5087 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |