Please login to your online account to display your discounted pricing

anti-PBEF/Visfatin/NAMPT, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223795SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223795SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. P43490
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Immunogen Amino acid 395-412 FKDPVADPNKRSKKGRLS of human NAMPT protein were used as the immunogen.
Primary or Secondary Primary
Gene ID (Entrez) 10135
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, horse, cow, rat and dog, 81% homologous in pufferfish and 71% homologous in zebrafish.
Format Antiserum
Applications Western Blotting
Quantity 0.025mL
Regulatory Status RUO
Antigen PBEF/Visfatin/NAMPT