Please login to your online account to display your discounted pricing

anti-PBEF/Visfatin/NAMPT, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223795

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223795

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P43490
Gene ID (Entrez) 10135
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen PBEF/Visfatin/NAMPT
Formulation 100μL of crude rabbit sera with 50% glycerol
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, horse, cow, rat and dog, 81% homologous in pufferfish and 71% homologous in zebrafish.
Applications Western Blotting
Regulatory Status RUO
Immunogen Amino acid 395-412 FKDPVADPNKRSKKGRLS of human NAMPT protein were used as the immunogen.
Format Antiserum
Host Species Rabbit
Quantity 0.1mL