Please login to your online account to display your discounted pricing

anti-PBEF/Visfatin/NAMPT, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223667SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223667SS

  • / Each
 

Description & Specifications

Specifications

Antigen PBEF/Visfatin/NAMPT
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Antisera
Formulation 100μL of rabbit sera with 50% glycerol
Gene Accession No. P43490
Host Species Rabbit
Immunogen A portion of amino acid 30-80 of human NAMPT/Visfatin was used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10135
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and chicken, and 94% homologous in horse, cow, mouse and rat and 83% homologous in zebrafish. The NAMPT protein has multiple variants of varying molecular weights including ∽55.5kDa (alternative variant a), 41.9kDa (alternative variant b), 35.6kDa alternative variant c), 28.5kDa (alternative variant d), and 26.8 (alternative variant e). NBP2-23795 detects an ∽38kDa band in western blot in the cells and tissues screened. This band may correspond to NAMPT, alternative variants b or c, or another NAMPT form.
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat