Please login to your online account to display your discounted pricing

anti-PBEF/Visfatin/NAMPT, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 30-80 of human NAMPT/Visfatin was used as the immunogen.
Antigen PBEF/Visfatin/NAMPT
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223667SS

 
novus biologicals canada
NBP223667SS
Unlabeled 0.025mL Each for $139.00

NBP223667

 
novus biologicals canada
NBP223667
Unlabeled 0.1mL Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 30-80 of human NAMPT/Visfatin was used as the immunogen.
Antigen PBEF/Visfatin/NAMPT
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and chicken, and 94% homologous in horse, cow, mouse and rat and 83% homologous in zebrafish. The NAMPT protein has multiple variants of varying molecular weights including ∽55.5kDa (alternative variant a), 41.9kDa (alternative variant b), 35.6 kDa alternative variant c), 28.5kDa (alternative variant d), and 26.8 (alternative variant e). NBP2-23795 detects an ∽38 kDa band in western blot in the cells and tissues screened. This band may correspond to NAMPT, alternative variants b or c, or another NAMPT form.
Gene Accession No. P43490
Gene ID (Entrez) 10135
Formulation 100μL of rabbit sera with 50% glycerol