Please login to your online account to display your discounted pricing

anti-Pax5/BSAP, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 1-29 of human Pax5. It will cross-react with mouse Pax5.
Antigen Pax5/BSAP
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224948SS

 
novus biologicals canada
NBP224948SS
Unlabeled 0.025mg Each for $109.00

NBP224948

 
novus biologicals canada
NBP224948
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 1-29 of human Pax5. It will cross-react with mouse Pax5.
Antigen Pax5/BSAP
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. Q02548
Gene ID (Entrez) 5079
Formulation Antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL