Please login to your online account to display your discounted pricing

anti-Pax5/BSAP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224948SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224948SS

  • / Each
 

Description & Specifications

Specifications

Antigen Pax5/BSAP
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation Antibody in 200μL PBS containing 0.2% gelatin
Gene Accession No. Q02548
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 1-29 of human Pax5. It will cross-react with mouse Pax5.
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5079
Target Species Human
Target Species Murine