Please login to your online account to display your discounted pricing

anti-pan Rho Polyclonal, Thermo Scientific™


pan Rho Polyclonal Ab OSC00025W from Thermo Scientific™ specifically detects pan Rho in Hu, Ms and Rt samples, and it is validated for IHC and WB

Manufacturer: pierce biotechnology inc  OSC00025W

 View more versions of this product

Catalog No. PIOSC00025W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Symbols RHOA, RHOB, RHOC, RHOF, RHOQ
Applications Immunohistochemistry
Applications Western Blot
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Gene Accession No. P08134, P17081, P61586, P61589, P62745, P62746, P62747, Q62159, Q8BYP3, Q8R527, Q9HBH0, Q9JJL4, Q9QUI0
Gene Alias Rho-related GTP-binding protein RhOA, RhOB, RhOC, RhOF, RhOQ, RAC1, RAC2, RAC3, CDC42
Species Reactivity Human, Mouse, Rat
Storage Requirements avoiding freeze/thaw cycles.
Gene ID (Entrez) 104215, 117273, 11848, 11852, 11853, 23433, 23912, 387, 388, 389, 54509, 64373, 85428
Quantity 100μL
Host Species Rabbit
Antigen pan Rho
Immunogen A synthetic peptide from human/rat/mouse/chicken/dog/frog
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled

pan Rho Polyclonal Ab OSC00025W from Thermo Scientific™ specifically detects pan Rho in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from human/rat/mouse/chicken/dog/frog
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Rho-related GTP-binding protein RhOA, RhOB, RhOC, RhOF, RhOQ, RAC1, RAC2, RAC3, CDC42
Uniprot Accession: P08134, P17081, P61586, P61589, P62745, P62746, P62747, Q62159, Q8BYP3, Q8R527, Q9HBH0, Q9JJL4, Q9QUI0
Entrez GeneID: 104215, 117273, 11848, 11852, 11853, 23433, 23912, 387, 388, 389, 54509, 64373, 85428
Gene Symbol: RHOA, RHOB, RHOC, RHOF, RHOQ
Glycerol (1:1) may be added for additional stability.