Please login to your online account to display your discounted pricing

anti-pan Rho Polyclonal, Thermo Scientific™

pan Rho Polyclonal Ab OSC00025W from Thermo Scientific™ specifically detects pan Rho in Hu, Ms and Rt samples, and it is validated for IHC and WB

$424.20 - $424.20

Specifications

Antigen pan Rho
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00025W

 
pierce biotechnology inc
OSC00025W
Unlabeled 100μL Each for $424.20

PIOSR00297G

 
pierce biotechnology inc
OSR00297G
Unlabeled 500μg Each for $424.20
Description & Specifications

Specifications

Antigen pan Rho
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide from human/rat/mouse/chicken/dog/frog
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

pan Rho Polyclonal Ab OSC00025W from Thermo Scientific™ specifically detects pan Rho in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from human/rat/mouse/chicken/dog/frog
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Rho-related GTP-binding protein RhOA, RhOB, RhOC, RhOF, RhOQ, RAC1, RAC2, RAC3, CDC42
Uniprot Accession: P08134, P17081, P61586, P61589, P62745, P62746, P62747, Q62159, Q8BYP3, Q8R527, Q9HBH0, Q9JJL4, Q9QUI0
Gene Symbol: RHOA
Entrez GeneID: 104215, 117273, 11848, 11852, 11853, 23433, 23912, 387, 388, 389, 54509, 64373, 85428
OSC00025W detects pan Rho in Human, Mouse and Rat samples. OSC00025W has been successfully used in Immunohistochemistry and Western Blot procedures. OSC00025W The immunogen is A synthetic peptide from human/rat/mouse/chicken/dog/frog.