Please login to your online account to display your discounted pricing

anti-PACS1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human PACS-1 was used as the immunogen.
Antigen PACS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224496

 
novus biologicals canada
NBP224496
Unlabeled 0.1mg Each for $257.41

NBP224496SS

 
novus biologicals canada
NBP224496SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human PACS-1 was used as the immunogen.
Antigen PACS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_e and CRA_f), horse, cow and chimpanzee, 94% homologous in mouse and rat, 89% homologous in opossum, platypus and dog and 78% homologous in chicken.
Gene Accession No. Q6VY07
Gene ID (Entrez) 55690
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL