missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-p73, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide CTHSPYAQPSS corresponding to amino acids 95-104 of human p73 was used as immunogen.
Antigen p73
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056435 Novus Biologicals Canada
NB10056435
0.1mg Each for $255.80
NB10056435T Novus Biologicals Canada
NB10056435SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
p73
Affinity Purified
RUO
This peptide sequence is 100% conserved across multiple p73 isoforms and between rat, mouse, dog and bovine p73. Researchers are encourage to use the the immunogen sequence information and available Bioinformatics data bases to obtain additional information regarding the amino acid sizes associated with various p73 isoforms. Cross reacts with Human. Predicted to react with Chicken, Chimpanzee, Dog, Mouse, New World Monkey and Rat.
7161
0.5mg/mL
A synthetic peptide CTHSPYAQPSS corresponding to amino acids 95-104 of human p73 was used as immunogen.
Polyclonal
Primary
Simple Western,Western Blot
NP_005418
PBS containing 0.2% gelatin