Please login to your online account to display your discounted pricing

anti-p73, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056435

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056435

  • / Each
 

Description & Specifications

Specifications

Applications Simple Western
Applications Western Blotting
Test Specificity This peptide sequence is 100% conserved across multiple p73 isoforms and between rat, mouse, dog and bovine p73. Researchers are encourage to use the the immunogen sequence information and available Bioinformatics data bases to obtain additional information regarding the amino acid sizes associated with various p73 isoforms. Cross reacts with Human. Predicted to react with Chicken, Chimpanzee, Dog, Mouse, New World Monkey and Rat.
Formulation PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Cross Reactivity Human
Gene Accession No. NP_005418
Gene ID (Entrez) 7161
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit
Immunogen A synthetic peptide CTHSPYAQPSS corresponding to amino acids 95-104 of human p73 was used as immunogen.
Primary or Secondary Primary
Antigen p73
Regulatory Status RUO
Quantity 0.1mg