Please login to your online account to display your discounted pricing

anti-p62/SQSTM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142822SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142822SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. Q13501
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8878
Concentration 1mg/mL
Antigen p62/SQSTM1
Quantity 0.025mL
Host Species Rabbit
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 400-450). [Swiss-Prot Q13501]
Format Affinity Purified