Please login to your online account to display your discounted pricing

anti-p38 beta/MAPK11, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to a portion amino acids 1-100 was used as immunogen
Antigen p38 beta/MAPK11
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056446

 
novus biologicals canada
NB10056446
Unlabeled 0.1mg Each for $255.80

NB10056446T

 
novus biologicals canada
NB10056446SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to a portion amino acids 1-100 was used as immunogen
Antigen p38 beta/MAPK11
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In human or mouse brain, a 38 kDa band is observed.
Gene Accession No. NP_002742
Gene ID (Entrez) 5600
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL