Please login to your online account to display your discounted pricing

anti-p35 baculovirus, Polyclonal, Novus Biologicals™

$144.63 - $307.00

Specifications

Host Species Rabbit
Immunogen Full-length recombinant p35 baculovirus protein was used as immunogen.
Antigen p35 baculovirus
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056153

 
novus biologicals canada
NB10056153
Unlabeled 0.05mL Each for $307.00

NB10056153T

 
novus biologicals canada
NB10056153SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full-length recombinant p35 baculovirus protein was used as immunogen.
Antigen p35 baculovirus
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity p35 baculovirus may be observed as one or multiple bands on western blot: p35 baculovirus is cleaved during its reaction with caspases. This results in cleavage of p35; 10 and 25 kDa fragments have been described. Additional breakdown p35 products may also result. Multiple specie cross reactivity. Not yet tested in other species.
Formulation Neat serum