Please login to your online account to display your discounted pricing

anti-P2Y2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11039032SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11039032T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Formulation PBS
Format Affinity Purified
Antigen P2Y2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Quantity 0.025mg
Gene ID (Entrez) 5029
Conjugate Unlabeled
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. P41231
Immunogen A synthetic peptide made to a C-terminal portion of the human P2Y2 protein (between animo acids 300-377) [UniProt P41231].
Host Species Rabbit