Please login to your online account to display your discounted pricing

anti-p19ARF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200106

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200106

  • / Each
 

Description & Specifications

Specifications

Antigen p19ARF
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.32mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 12578
Test Specificity Mouse. Immunogen sequence has 93% homology to rat, 86% homology to porcine and 80% homology to human.
Target Species Murine