Please login to your online account to display your discounted pricing

anti-p16INK4a / CDKN2A/p14ARF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200158SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200158SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Gene ID (Entrez) 1029
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Gene Accession No. NP_478102.1
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Antigen p16INK4a / CDKN2A/p14ARF
Format Affinity Purified
Immunogen The epitope recognized by this antibody maps to a region between residue 75 and 125 of human p14ARF using the numbering given in entry NP_478102.1 (GeneID 1029).
Regulatory Status RUO